VWR® Ergonomic Smooth Floor Mats, Gray
Proveedor: VWR
VWR®
INS0306-S INS0305-S INS0209-S INS0320-S INS0410-S INS0204-S INS0207-S INS0208-S INS0310-S INS0206-S INS0203-S INS0308-S INS0203SILVWR INS0206SILVWR INS0307-S INS0204SILVWR INS0412-S INS0205SILVWR INS0210-S INS0407-S INS0304-S INS0205-S INS0303-S INS0408-S INS0420-S INS0220-S INS0304SILVWR INS0405-S INS0406-S
89185-264EA 5386.97 MXN
89185-228 89185-306 89185-308 89185-268 CA89185-234 89185-302 89185-304 89185-242 89185-264 CA89185-230 89185-266 CA89185-232 76319-000 CA89185-228 89185-312 CA89185-266 89185-236 89185-238 89185-232 89185-278 89185-234 89185-272 89185-230 89185-274 89185-270 76318-992 76318-994 76318-996 76318-998
VWR® Ergonomic Smooth Floor Mats, Gray
Alfombrillas Alfombrillas antifatiga
The VWR® Smooth Mat is a true anti-fatigue mat. This smooth mat has the comfort of a polyurethane mat and the added benefit of easy clean-ability due to its closed-cell/non-porous top surface.
  • Stimulate the body to increase blood circulation and alterness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Antistatic and meets ISO Class 5 (Class 100) compliance
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up, eliminating tripping hazards. With the VWR Smooth mat, you will never have overlapping or worn-out connecting mats, also eliminating tripping hazards.
Constructed of the highest quality using only virgin raw material, this mat has lifetime ergonomic benefits. The VWR® Smooth mat is silicone & latex-free. Using its unique ergonomic designs, this smooth mat prevents common problems encountered in standing environments while stimulating the body to increase blood circulation and alertness that will reduce sick leave, workers’ compensation and quality issues. The VWR® Smooth mat is a solid mat throughout.
It also can be customized to any size and any shape, always in one piece.
Certificaciones: Meets Cleanroom ISO Class 5 (Class 100) compliance.