VWR® Ergonomic Bubble Floor Mats, Blue
Proveedor: Avantor
VWR®
IN0213-BLU IN0415-BLU IN0209-BLU IN0404-BLU IN0318-BLU IN0314-BLU IN0416-BLU IN0305BLUVWR IN0320-BLU IN0410-BLU IN0316-BLU IN0203BLUVWR IN0411-BLU IN0313-BLU IN0420-BLU IN0215-BLU IN0217-BLU IN0317-BLU IN0405BLUVWR IN0205-BLU IN0214-BLU IN0417-BLU IN0206BLUVWR IN0419-BLU IN0309-BLU IN0405-BLU IN0414-BLU IN0315-BLU IN0319-BLU IN0211-BLU IN0204-BLU IN0220-BLU IN0406-BLU IN0218-BLU IN0408-BLU IN0407-BLU IN0307-BLU IN0207BLUVWR IN0206-BLU IN0303-BLU IN0310-BLU IN0409-BLU IN0418-BLU IN0304-BLU IN0412-BLU IN0210-BLU IN0305-BLU IN0311-BLU IN0208-BLU IN0207-BLU IN0216-BLU IN0306-BLU IN0308-BLU IN0203-BLU IN0212-BLU IN0219-BLU IN0312-BLU
89184-914EA 5985.34 MXN
76318-890 76318-892 76318-846 76318-848 89184-924 89184-922 89184-920 76318-882 76318-884 89184-960 76318-886 76318-842 76318-888 76318-844 CA89184-918 76318-858 89184-918 89184-916 89184-914 89184-958 89184-912 89184-956 76318-894 76318-850 89184-910 76318-896 76318-852 89184-952 76318-854 76318-898 89184-950 76318-856 76319-118 76319-116 76319-114 76318-870 76319-112 76318-902 76318-868 CA89184-916 76318-904 89184-948 89184-946 CA89184-910 76318-860 89184-984 CA89184-950 76318-862 76318-864 76318-900 76318-866 76318-880 CA89184-984 76318-872 76318-874 76318-876 76318-878
VWR® Ergonomic Bubble Floor Mats, Blue
Alfombrillas Alfombrillas antifatiga
The VWR® Bubble Mat is a true ergonomic mat. The VWR Bubble mat has the comfort of a polyurethane mat and the added benefit of easy clean-ability due to its closed-cell/non-porous top surface.
  • Stimulate the body to increase blood circulation and alertness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Antistatic and meets ISO Class 5 (Class 100) compliance
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up, eliminating tripping hazards. With the VWR® Bubble mat, you will never have overlapping or worn-out connecting mats, which also eliminates tripping hazards.
Constructed of the highest quality using only virgin raw material, this bubble mat has lifetime ergonomic benefits. The VWR Bubble mat is silicone & latex-free. Using its unique ergonomic designs, this mat prevents common problems encountered in standing environments, while stimulating the body to increase blood circulation and alertness that will reduce sick leave, workers' compensation and quality issues. The VWR® Bubble mat is a solid mat throughout, which means no hollow bubbles that collapse.
The VWR® Bubble mat can be customized to any size and any shape, always in one piece.
Certificaciones: Meets Cleanroom ISO Class 5 (Class 100) compliance.