VWR® Ergonomic Economy Smooth Floor Mats, Black
Proveedor: Avantor
SB0204 SB0404 SB0206 SB0205 SB0306VWR SB0208VWR SB0207VWR SB0209 SB0305VWR SB0307 SB0305 SB0203 SB0308VWR SB0407 SB0308 SB0408 SB0208 SB0207 SB0410 SB0405 SB0306 SB0203VWR SB0309 SB0303 SB0206VWR SB0204VWR SB0310 SB0304 SB0304VWR SB0406 SB0320
89185-446EA 3725.45 MXN
89185-448 89185-444 89185-488 89185-446 89185-524 CA89185-478 89185-484 89185-440 CA89185-450 89185-442 89185-486 89185-520 89185-480 89185-482 89185-518 89185-514 CA89185-446 CA89185-448 89185-516 CA89185-486 CA89185-442 89185-510 89185-478 89185-512 CA89185-482 89185-452 CA89185-440 89185-476 CA89185-480 89185-450 89185-490
VWR® Ergonomic Economy Smooth Floor Mats, Black
Alfombrillas Alfombrillas antifatiga
The VWR® Economy Smooth Mat is a low-density polyurethane material with an anti-fatigue profile. This flat mat is ideal for dry, non-critical environments combining both sitting and standing work areas. No smooth matting available delivers as much relief and comfort at this price. The VWR® Economy Smooth Mat can be customized to any size and any shape, always in one piece.
  • Stimulate the body to increase blood circulation and alertness
  • Molded beveled edges eliminate tripping hazards
  • Silcone free, latex free
  • Easy-to-clean, closed-cell/nonporous top surface
  • Highly resistant to chemicals and UV light
Safety features include molded beveled edges that will never curl up, eliminating tripping hazards
The VWR® Economy Smooth Mat has lifetime ergonomic benefits. Using its unique ergonomic designs, this flat mat prevents common problems encountered in standing environments while stimulating the body to increase blood circulation and alertness that will reduce sick leave, workers' compensation and quality issues. The VWR® Economy Smooth Mat is a solid mat throughout. This mat is silicone & latex-free.
With the VWR® Economy Smooth Mat, you will never have overlapping or worn-out connecting mats, which also eliminates tripping hazards.