VWR® Advanced round top hotplate-stirrers deliver accurate and repeatable results.
- Excellent temperature uniformity
- Industry Leading Safety Features
- Cool touch, chemical-resistant housing
- LED display for temperature
- Accessories easily convert unit to a multi-sample reaction station
- Now with Best-in-Class, Exclusive 5-Year Warranty
These units are microprocessor controlled and have an Easy-to-read LED display for set-point temperature
Top plate has a 135mm (5.3") diameter, and is made of durable stainless steel that will not crack. A ramping feature that gradually increases speed results in improved safety, enhanced coupling, and excellent low end speed control. A built-in memory recall provides users with the last set temperature, even after the unit has been turned off. For additional safety, a hot symbol warning light is illuminated when the hot plate is in use, and remains lit until the top plate is sufficiently cooled.
Enhanced electronics regulate the robust heater, which brings samples to the desired temperature quickly and efficiently. Temperature measurement is controlled more accurately by utilizing an internal RTD. Speed is precisely controlled, and provides consistent stirring at all speeds. The low-profile design utilizes minimal bench space and permits use within a fume hood. Unit housing is constructed of a heat- and chemical-resistant polymer that remains cool to the touch during operation. The spill-resistant design channels fluids away from internal components in the event of spills. Units are equipped with redundant safety controls, which will shut down unit in case of heater malfunction.
Control panel features easy-to-use controls which allow users to dial in adjustments. Rear housing features an integral support rod holder with locking knob to accept the optional Support Rod and Clamp Kit.
Certificaciones: RoHS certified. TUV certified.
Información para pedidos: Unit includes a 234 cm (92") detachable, 3-wire cord and plug, and a 3.8 cm (11/2") PTFE coated stir bar.