Nunc® CryoPens, Thermo Scientific
Proveedor: Thermo Scientific
343850 343850
CA24270-010 24270-011
Nunc® CryoPens, Thermo Scientific
Bolígrafos Rotuladores criogénicos
Nunc® CryoPens come with indelible, quick-drying ink and extra fine tips
They are ideal for use with Nunc® Cryo Color Coders as part of a versatile and comprehensive filing system.