VWR® White Light Transilluminators
Proveedor: VWR
95-0209-01
89131-472EA 18713.75 MXN
89131-472
VWR® White Light Transilluminators
Transiluminadores Transiluminadores de luz blanca
Transilluminators provide brilliant back illumination of transparent materials that are placed on the filter area.
  • Brilliant backlit illumination
  • Designed to provide increased UV sensitivity and uniformity
  • Reduced electrical consumption
  • Durable Plexiglass and glass filter combination
The transilluminators provide a highly sensitive method to detect double-stranded nucleic acids that have been labeled with fluorescent dyes such as ethidium bromide or acridine orange
Compact white light transilluminators are used for viewing Coomassie blue or silver stained gels. Uniform white light diffusion enables viewing of autoradiographs, X-ray film, film positives, negatives, and microtiterplates. Plexiglass and glass filter combination are sealed into the unit for durability.
The transilluminators are uniquely designed with increased UV intensity and uniformity, instant on capabilities, no lamp flicker and reduced electrical consumption.