VWR® White Light/UV Transilluminators
Proveedor: VWR
95-0554-01
89131-476EA 67111.39 MXN
89131-476
VWR® White Light/UV Transilluminators
Transiluminadores Transiluminadores de luz blanca UV
Transilluminators provide brilliant back illumination of transparent materials that are placed on the filter area.
  • Provide brilliant back illumination of transparent materials
  • Highly sensitive method to detect dyed double-stranded nucleic acids
  • Supply UV and white light side by side
The transilluminators provide a highly sensitive method to detect double-stranded nucleic acids that have been labeled with fluorescent dyes such as ethidium bromide or acridine orange
Large format White/UV transilluminators supply 302nm UV and white light side by side. These transilluminators enable researchers to maximize their space and application capabilities by viewing fluorescent gels on the UV side or viewing Coomassie blue or silver stained gels on the white light side. Easily switch between lighting with the easy access knob.
Single stranded nucleic acids may be detected, but with a lower excitation 302nm UV wavelength more sensitive for nucleic acid visualization than the 365nm model. The transilluminators are uniquely designed with increased UV intensity and uniformity, instant on capabilities, no lamp flicker and reduced electrical consumption.