Standard Kel-F Hub Luer Lock Needles, Hamilton Company
Proveedor: Hamilton
7746-01 7746-06 7746-04 90151 7746-03 90118 7746-05 90130 90153 90134 7746-10 90111 7746-08 7746-09 90115 90113 7746-11 7746-12 90120 90123 90117 90116 7746-16 90127 7746-15 90121 90129 7746-07 90119 90110 7746-13 7746-02 90152 90114 90112
60379-245PK 1913.88 MXN
82010-382 89220-936 89220-934 89221-160 89220-932 89220-930 82010-400 82010-402 82010-386 89221-166 72310-220 60379-245 89221-162 72310-202 72310-224 89221-164 89220-928 82010-392 72310-206 82010-394 72310-226 89221-172 82010-390 82010-378 82010-396 82010-404 72310-210 72310-232 89221-178 72310-230 72310-214 72310-236 89221-174 72310-234 89221-176
Standard Kel-F Hub Luer Lock Needles, Hamilton Company
Agujas de jeringas
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry.
Each is manufactured to achieve the highest level of accuracy and precision possible. Syringe Terminations: Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
PTFE Luer Lock (TLL)- This termination has a PTFE, male luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters and dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10. Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10. Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10.