Nunc® MicroWell™ 96-Well Optical-Bottom Plates, Thermo Scientific
Proveedor: Thermo Scientific
160376 164588 164590 164590 265300 164588 265300
73520-168CS 48416.47 MXN
CA73520-750 CA73520-178 73520-168 73520-178 CA73520-168 CA73520-174 73520-174
Nunc® MicroWell™ 96-Well Optical-Bottom Plates, Thermo Scientific
Microplacas Microplacas para inmunología
The Plates are optimized for fluorescence and luminescence in cell culture and microscopic applications
The coverglass bottom has minimum light scatter and low autofluorescence, ensuring accurate results due to higher signal-to-noise ratio.