Open Combination Kjeldahl Digestion/Distillation Apparatus, Labconco®
Proveedor: Labconco
2123215 2123313 2123315 2123211 2123312 2123212 2123213
10129-214EA 833447.16 MXN
10129-210 10129-220 10129-212 10129-222 10129-218 10129-214 10129-216
Open Combination Kjeldahl Digestion/Distillation Apparatus, Labconco®
Digestores Estaciones de trabajo de mineralización
Open Combination Kjeldahl Digestion/Distillation Apparatus
Combines macro digestion and distillation processes for nitrogen determination of samples into one space-saving unit. Able to digest and distill six or twelve samples at one time. Open Combination Kjeldahl units are supported by a sturdy, tubular steel framework, with the distillation apparatus mounted above the digestion portion.
Información para pedidos: Requires PVC Remote Acid Exhaust Blower, ductwork, and 500 or 800 ml Kjeldahl flasks (not included). Six- and Twelve-Place units are available.