Economical Stainless Steel Weight Sets, Troemner
Proveedor: Troemner
30390963 30390953 30390956 30390974 30390958 30390966 30390970 30390968 30390967 30390957 30390973
97044-264EA 3920.25 MXN
97044-276 97044-266 97044-274 97044-264 97044-272 97044-262 97044-270 97044-282 97044-280 97044-278 97044-268
Economical Stainless Steel Weight Sets, Troemner
Pesas Pesas de calibración
The makeup of Economical Stainless Steel Weight Sets varies.
Weights 1 g and above are carefully manufactured of stainless Steel and are cylindrical in shape for easy gripping. Weights 1 g to 100 g are of one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight. Weights 200 g through 2 kg have an adjusting cavity on the top that is sealed with a plug. Economical Stainless Steel weights 1 g and above are perfectly flat on top to stack easily on center. Weights 500 mg and lower are made of sheet metal and are one-piece construction with one side turned up to make them easy to handle with forceps. 500 mg and 200 mg weights are made of stainless steel while weights 100 mg and below are made of aluminum.
Certificaciones: Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.