Dropper Bottles, Clear, Round, Qorpak
Proveedor: Qorpak
GLC-05726 GLC-05724 GLC-05723
70000-006CS 2382.07 MXN
70000-002 70000-006 70000-008
Dropper Bottles, Clear, Round, Qorpak
Frascos Frascos de goteo
Round Dropper Bottles are supplied with attached glass dropper assemblies.
Dropper Bottles are ideal for dispensing small amounts of liquid. They feature black polypropylene closures, rubber bulbs and straight point glass dropper tubes. Available in Amber or Clear glass.