Packer Bottles, PET, Clear, Wide Mouth, Qorpak
Proveedor: Qorpak
PLC-07300 PLC-06762 PLC-06836 274632 PLC-08925 PLC-08822 PLA-06570 PLC-07520 PLC-06838 PLC-09008 PLC-06837 PLC-06877 PLC-11297 PLC-07097 PLC-06996 PLC-06594
11311-640CS 6654.84 MXN
75880-446 11311-636 11311-640 89234-738 82020-058 76487-252 76487-362 76487-250 11311-639 76487-360 76487-256 76487-366 76487-254 76487-364 14003-560 30617-180
Packer Bottles, PET, Clear, Wide Mouth, Qorpak
Frascos Frascos Roller
PET Packer Bottles feature glass like clarity and are durable and rigid.
  • PET packers are stress and impact resistant
  • PET bottles are ideal for oil analysis
They have exceptional gas and moisture barrier properties and a good alcohol and solvent barrier.
Información para pedidos: Bottle 30617-180 is Vacuum and Ionized. Bottle 89234-738 is Super Cleaned for Oil Analysis.