VWR® General Purpose Chromatography Refrigerator
MH45SS-GAEE-VW MH49SS-GAEE-VW MH30SS-GARE-VW MH72SS-GAEE-VW MH45PA-GAEE-VW MH72SS-GARE-VW MH72SS-GARE-VW MH72PA-GAEE-VW MH45PA-GARE-VW MH30PA-GAEE-VW MH45SS-GAEE-VW MH30PA-GAEE-VW MH49PA-GAEE-VW MH72PA-GARE-VW MH30PA-GARE-VW MH38PA-GAEE-VW MH49PA-GARE-VW MH45SS-GARE-VW MH45SS-GARE-VW MH49SS-GARE-VW MH30SS-GAEE-VW MH72SS-GAEE-VW MH72PA-GARE-VW MH45PA-GARE-VW MH30PA-GARE-VW MH49PA-GARE-VW MH45PA-GAEE-VW MH38PA-GARE-VW MH30SS-GARE-VW MH38PA-GAEE-VW MH49PA-GAEE-VW MH72PA-GAEE-VW MH49SS-GARE-VW MH49SS-GAEE-VW MH30SS-GAEE-VW MH38PA-GARE-VW
89209-812EA 278474.19 MXN
CA89209-842 CA89209-820 89209-830 CA89209-844 CA89209-822 89209-812 89209-834 89209-832 CA89209-840 CA89209-828 89209-838 89209-816 89209-814 89209-836 CA89209-846 CA89209-824 CA89209-826 89209-818 CA89209-830 89209-842 89209-820 89209-840 CA89209-832 89209-824 89209-846 89209-844 89209-822 89209-828 CA89209-838 CA89209-816 CA89209-818 89209-826 CA89209-812 CA89209-834 CA89209-836 CA89209-814
VWR® General Purpose Chromatography Refrigerator
VWR® Laboratory Chromatography Refrigerators are designed for general-purpose applications in laboratory environments, and to meet the rigorous demands life science research.
  • Adjustable temperature range of 1 to 12°C (34 to 54°F)
  • 2.5 cm (1") rear access port
  • Two 5 cm (2") side access ports
Standard features include a dual-pane glass door, one internal duplex outlet, set of four casters, bright easy-to-read digital display, and a full set of audio and visual alarms including door ajar, high/low temperatures, and alarm test.
Certificaciones: UL/C-UL Listed.
Información para pedidos: Units come with wheels and installation is required. All VWR® General-Purpose Chromatography Refrigerators are supplied with a two-year parts and labor warranty.