Replacement Bag for Bag-in/Bag Out HEPA Filter, Labconco®
Proveedor: Labconco
3776025 3776024 3776023
82018-216EA 8233.85 MXN
82018-214 82018-216 82018-218
Replacement Bag for Bag-in/Bag Out HEPA Filter, Labconco®
Cajas de guantes
Replacement bag for a true bag-in/bag-out HEPA filter system
This bag includes a sealed bag with gloves allowing a technician to remove a contaminated filter without exposing the filter or contaminated plenum to personnel or the environment. HEPA filter replacement should only be performed by a certified technician. For use with designated size XPert Systems and Stations.