Liebig Condenser, 'No Hold Up', Ace Glass Incorporated
Proveedor: Ace Glass
5998-15 5998-24 5998-13 5998-25 5998-14 5998-12 5998-23 5998-04 5998-16 5998-17 5998-22
89053-934EA 4439.07 MXN
89053-946 89053-934 89053-944 89053-998 89053-928 89053-938 89053-948 89053-870 89053-898 89053-942 89053-940
Liebig Condenser, 'No Hold Up', Ace Glass Incorporated
Condensadores
Liebig condenser, "No Hold Up", with standard taper reinforced outer joint at top and standard taper inner joint at bottom
Fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections.