Agarose, Molecular Biology Grade, IBI Scientific
Proveedor: IBI Scientific
IB70035 IB70042 IB70040 IB70041 IB70047 IB70045 IB70046 IB70042
IB70040EA 4679.26 MXN
87004-980 IB70042 95043-304 97060-748 95000-010 IB70040 490007-708 97060-746
Agarose, Molecular Biology Grade, IBI Scientific
Geles de eletroforesis Agarosa y acrilamida
IBI Molecular Biology agarose products are highly refined and highly purified
Some features of IBI agarose include extraordinary mechanical resistance which insures safe and easy handling, exceptionally low absorption of staining reagents, excellent transparency which allows for high visibility, and greater thermal stability due to high hysteresis.
IBI agarose can be used for Nucleic Acid Electrophoresis, Analytical Electrophoresis, Preparative Electrophoresis, Blotting, and Protein Electrophoresis as in Radial Immunodiffusion.
Each lot is physically tested and verified for performance in its' designated application. IBI applies strict testing parameters to insure lot to lot consistency. IBI agarose products are offered in a wide variety of package sizes to fit the needs of any research. Each bottle of IBI agarose comes packaged with a specification sheet, preparation instructions, and MSDS for your convenience.