Nalgene® Heavy Duty Cylindrical Tanks with Cover, LLDPE, Thermo Scientific
Proveedor: Thermo Scientific
11100-0080 11100-0007 11100-0015 11100-0100 11100-0200 11100-0030 11100-0005 11100-0010 11100-0150 11100-0055
60463-160EA 24837.12 MXN
60463-160 60463-150 60462-060 60464-010 60463-156 60462-048 60462-026 60463-158 60463-152 60463-154
Nalgene® Heavy Duty Cylindrical Tanks with Cover, LLDPE, Thermo Scientific
Depósitos Depósitos cilíndricos
Nalgene Heavy Duty Cylindrical Tanks with cover are made of Linear Low-Density Polyethylene
They are rigid, hard surfaced and offered in sizes ranging from 19 L to 757 L. Factory Spigots can be installed on tanks up to 378 L.