Nalgene® PP Cylindrical Tank with Cove, Thermo Scientific
Proveedor: Thermo Scientific
11200-0007 11200-0015 11200-0055 11200-0100 11200-0010 11200-0030 11200-0005
60455-190EA 27033.6 MXN
60455-186 60455-176 60455-178 60455-182 60455-184 60455-190 60455-180
Nalgene® PP Cylindrical Tank with Cove, Thermo Scientific
Depósitos Depósitos cilíndricos
Nalgene Polypropylene Tanks with covers provide good resistance to stress cracking and are tolerant of many organic chemicals
Tanks are available in sizes ranging from 19 L to 378 L, and can be modified with factory-installed spigots.