Refine™ Protective Eyewear, 3M™
Proveedor: 3M
Refine™
7000127526
89136-098CS 5904.99 MXN
89136-098
Refine™ Protective Eyewear, 3M™
Gafas
Lightweight, slim frame is specially sized for the slim profiles of women.
  • Lightweight, slim-frame fit adjusts comfortably to female facial profile
  • Antifog coating and polycarbonate lens deliver clarity of vision
Goggles offer better coverage with fewer gaps to help keep out particulates and debris.
Certificaciones: Meets ANSI Z87.1 requirements.