Four Star™ Heat Resistant Brushes, EXCELTA®
Proveedor: Excelta Corporation
211A-N 211C-N 211D-N 211B-N
94024-172EA 604.76 MXN
94024-174 94024-176 94024-178 94024-172
Four Star™ Heat Resistant Brushes, EXCELTA®
Cepillos
The Nylon fibers are epoxy set into the Delrin handles with a high temperature adhesive to resist deterioration in an autoclave.
  • Impervious to solvents and common chemicals, as well as bacteria
  • Heat resistant to 250°F (121°C) and electrically insulative
The soft non shedding Nylon fibers are perfect for working on delicate parts. Brushes should be cleaned prior to cleanroom use.