Bushings, Ace Glass
Proveedor: Ace Glass
7506-12 7506-35 7506-01 7506-02 7506-06 7506-08 7506-14 7506-15 7506-31 7506-23 7506-09 7506-29 7506-10 7506-11 7506-20 7506-27 7506-05 7506-03 7506-39 7506-33 7506-127
89059-226EA 845.14 MXN
89059-218 89059-228 89059-238 89059-216 89059-226 89083-162 89059-248 89059-214 89059-236 89059-246 89059-224 89083-164 89059-212 89059-234 89059-222 89059-244 89059-232 89059-220 89059-242 89059-230 89059-240
Bushings, Ace Glass
Racores y conectores
Front-seal bushing connector for use with Ace-Thred joints, threaded glass, or stainless steel connectors
Used to join threaded end to a reduced-end tube. Available in either nylon or PTFE.