Adapter, Stainless Steel, Circulator Hose, Ace Glass Incorporated
Proveedor: Ace Glass
12187-07 12187-05 12187-10 12187-14 12187-12
89097-264EA 5082.76 MXN
71001-582 71001-584 89097-266 89097-264 71001-580
Adapter, Stainless Steel, Circulator Hose, Ace Glass Incorporated
Racores y conectores
These stainless steel adapters are designed for use with ACE jacketed glass pilot plant reactors and circulator hoses for popular circulating chillers.
  • Bottom spherical joint
  • Top M style male
  • Stainless steel
This flask-to-hose adapter connects the o-ring ball joints on the jacket inlet/outlet to hoses to/from the circulator.