Bottle, Specific Gravity Calibrated, Ace Glass Incorporated
Proveedor: Ace Glass
5425-10 5425-20 5425-05 5425-15
89051-714EA 6337.84 MXN
89051-708 89051-714 89051-712 89051-710
Bottle, Specific Gravity Calibrated, Ace Glass Incorporated
Botellas con gravedad específica
This Guy-Lussac type bottle is calibrated (2mL: ±0.05L; 5, 10, and 25mL: ±0.10mL) and is engraved
Suitable for ASTM D 369. Comes complete with glass stopper. Fabricated from borosilicate glass.