Amber Glass Dropping Bottles
Proveedor: EMPIRE/EMCO
01A010A 01A002A W/ 405-5D-24-GR
470188-928EA 30.49 MXN
470189-418 470188-928 470188-926
Amber Glass Dropping Bottles
Frascos Frascos de goteo
Ideal for Small Quantities of Light Sensitive Chemicals.
  • Store stock reagents and solutions
  • Matching pipet assemblies are available separately