Cardboard Slide Box for 25 Slides
SBC25PETRO
470022-428CS 31147.58 MXN
470022-428
Cardboard Slide Box for 25 Slides
Archivadores para portaobjetos
Economical Storage and Transport of Slides.
  • Keep slides collections clean and safe
  • Economical option for slide storage
Store up to twenty-five 27×46 mm petrographic slides. Includes a numbered index insert.