Chromatography Paper Strips
Catalog # 470004-492
Supplier: PRECISION LABORATORIES SE
Specifications
- Description:Chromatography Paper Strips, Pack of 50
- Size:6 x ¾"
- Cat. no.:470004-492
- Supplier No.:CHROM-50-6X75
Specifications
About this item
Perfect for your paper chromatography needs.
- Package of 50
This chromatography paper is supplied in strips rather than rolls that are apt to stretch in order to assure the most even distribution of material being separated for more accurate results.