Clostridium tetani Slide
470180-896
: Avantor
- Description:Clostridium tetani,gram (+)
- Cat. no.:470180-896
- Stain:Gram (+)
- Supplier No.:WARD470180-896
- Orientation:Smear
Pathogenic bacteria
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods. Spores round and terminal. Agent of tetanus in humans.