Ward's® Cornell Unit Pinning Trays
Proveedor: CHARLES W. KANTY
Ward's®
302120 PINTRAY4X1 302100
470177-240EA 892.4 MXN
470178-304 470177-240 470306-678
Ward's® Cornell Unit Pinning Trays
Bandejas/Platos/Ollas Bandejas/Placas de laboratorio
Perfect for conditions where removing or regrouping specimens is frequent.
When you rearrange your specimens, you'll appreciate the convenience of these modular trays. Crafted of sturdy chipboard coated with white, enamel paper and an ethafoam pinning bottom that accepts pins easily and holds them securely.
Información para pedidos: All measurements are outside dimensions.