Low Form Porcelain Crucibles
Proveedor: SILICO & CHEMICO PORCELAIN SE
J236 J236 J243
470149-150CS 963.55 MXN
470149-150 470149-152 470200-314
Low Form Porcelain Crucibles
Crisoles
These low, wide crucibles are made from highest quality porcelain. All surfaces are glazed except the outside bottom and rim. Without covers.