Disposable Trays, Foam Fabricators
Proveedor: FOAM FABRICATORS, LTD.
BLANK
470149-646PK 416.22 MXN
470149-646
Disposable Trays, Foam Fabricators
Bandejas/Platos/Ollas Bandejas/Placas de laboratorio
These economical and disposable trays are well suited for beginning dissectors.
The wider base of these stackable styrofoam trays keeps them stable, avoiding accidental spills, and the thick bottom acts as a liner. Ideal size for small dissections life Earthworms, Clams , and Locust.