DNA on a Stick Lab Activity
Proveedor: Lab Activity
DD-115
470003-024EA 1551.55 MXN
470003-024
DNA on a Stick Lab Activity
Kits y actividades educativas para el aula
Discover the Appearance of DNA
  • Isolate you own DNA
  • DNA can be stained or run on a gel
An intriguing introduction to genetics studies, DNA On a Stick allows students to spool chromosomal DNA from solution onto a stick. Once students have collected the coiled DNA, it can be stained for further visualization or prepared for agarose gel electrophoresis.