Economy Polypropylene Low Form Beakers
Proveedor: MARYLAND PLASTICS INC SE
L-1230 L-1250 L-1270 L-1260 L-1220 L-1250 L-1260
470149-709CS 1143.45 MXN
470149-709 470153-379 470153-385 470149-924 470153-386 470206-318 470153-382
Economy Polypropylene Low Form Beakers
Vasos de precipitados
Ideal for school and kitting use.
  • Griffin Low Form Beakers
  • Inexpensive and Resistant
These affordable polypropylene beakers are unaffected by most acids, alkalis and solvents. The manufacturer describes them as disposable, but most teachers find them tough enough to reuse several times. Autoclavable to 121°C. Designed with a dripless pouring spout and graduations accurate to ±5%.