Fan Cart
Proveedor: SCIENCE INTERACTIVE GROUP
10-105TSS 10-105/01
470105-306EA 4539.35 MXN
470105-306 470223-112
Fan Cart
Materiales educativos Materiales Educativos de Física Mechanics Learning Activities
Produce constant acceleration with this fan cart.
  • Fan provides constant force, resulting in constant acceleration
  • Removable plate vividly demonstrates Newton's third law
Graphically illustrate uniform acceleration independent of gravity. A motor-driven propeller provides thrust to accelerate the cart along a smooth level surface.
Made of durable, lightweight plastic, the cart features a 1.5×12.5 cm propeller, four low friction wheels, and a 14×15 cm removable sail.
Powered by four AA batteries, purchased separately. Overall size: 11×19.5×11 cm.