Griffin Tubing Glass Cutter
Proveedor: HUMBOLDT MFG CO
H-13520
470122-404EA 574.77 MXN
470122-404
Griffin Tubing Glass Cutter
Cortadores
Hardened steel wheel cuts glass tubes up to 11/2" dia. Two V guides align the tubing and wheel is spring loaded to stay open.
  • Made of nickel-plated zinc-alloy die-casting