Heat and Temperature Set
Proveedor: Avantor
WARD470152-862
470152-862EA 10366.77 MXN
470152-862
Heat and Temperature Set
Kits y actividades educativas para el aula
Six Different Exercises Show the Effects of Heat Energy
  • Best Seller
  • Perform Many Experiments with this Set
A variety of investigations explore conduction, convection, radiation, and specific heat. The kit includes apparatus for examining thermal expansion, expansion rates, vapor pressure, gas convection, radiant heat, and specific heat to demonstrate the effects of heat energy on solids, liquids, and gases, as well as instructions for each demonstration. Smoke paper for gas convection experiment, required, is available separately.