Heavy Duty Steel Support Rods
Proveedor: VWR International
WLS78379-D WLS78379-A WLS78379-B WLS78379-G WLS78379-E WLS78379-C WLS78379-N WLS78379-H WLS78379-F
470123-502EA 217 MXN
470123-502 470123-500 470123-498 470123-496 470123-494 470123-492 470123-508 470123-490 470123-504
Heavy Duty Steel Support Rods
Varillas de retorta
Made of smooth finished steel, heavily nickel plated and highly polished. These support rods are squarely machined to fit bases accurately.
Opposite ends are rounded. For use with most bases and rod support clamps.