Order Entry
Hooke’s Law University Apparatus
Hooke’s Law University Apparatus
  470002-922
undefined
Hooke’s Law University Apparatus
  470002-922
$453.07
  Each (1UN)
  • Description:
    Hook's Law Apparatus, University
  • Cat. no.:
    470002-922
  • Supplier No.:
    PH0322A
This apparatus verifies that that the amount by which a material body is deformed (the strain) is linearly related to the force causing the deformation (the stress).
  • Durable Metal Base
  • Mirrored Scale
  • Slotted Weights
  • For Verifying Concepts Of Hooke's Law
This instrument is designed for advanced analysis of Hooke’s law and features a mirrored scale.
Constructed of a metal base with a 12 cm mirror scale, a spring is supported with a weight holder and 250 g slotted weights.