Boreal Science Student Stereomicroscopes
Proveedor: MOTIC HONG KONG LIMITED
Boreal
5540400 5572803 2/CS 5744900 S-20-P
470012-230EA 9510.98 MXN
470012-230 470145-720 470145-578 470145-710 470145-564
Boreal Science Student Stereomicroscopes
Microscopios
These stereomicroscopes have all the quality features and versatility of larger stereo microscopes in a compact size.
  • Adjustable Interpupillary Distance
  • Interchangeable Stage Plates: Translucent and Black/White
  • Lifetime Manufacturer's Warranty
  • Diopter Adjustment on One Eyetube
Features include 45° inclined viewing heads, post mounts that adjust the head up or down to accommodate larger objects on stage, and an easy-adjust collar for precise control of focus tension.
Rugged, die-cast metal construction and a reagent resistant finish make these stereomicroscopes durable.
Información para pedidos: This microscope includes removable soft eyecups and vinyl dust cover.