Order Entry
Ward's® Introductory Mineral Collection
  470025-258
 :  Avantor
undefined
Ward's® Introductory Mineral Collection
  470025-258
 :  Avantor
$1,004.40
  Each (1UN)
  • Description:
    Introductory Mineral Collection
  • No. of Samples:
    12
  • Cat. no.:
    470025-258
  • Supplier No.:
    WARD470025-258
Beginner’s Collection Illustrates Important Physical Properties
  • Cut by WARD's Geologists
  • Highest Quality Specimens
  • Great for Introducing Students to Minerals
Color, hardness, luster, cleavage, streak, common habit, and specific gravity are all represented in one compact, economical set of 12 minerals. Beginning geology students will easily recognize each of the wide range of physical properties when they examine the basic minerals. The set is housed in a compartmented collection box, and an identification list is included.
Contents:12 numbered samples, 1" x 2"Minerals:apatite, calcite, graphite, halite, hematite, limonite, microcline, muscovite, olivine, pyrite, quartz, selenite