LaMotte® Dissolved Oxygen Sampler
1054-DO
470021-298EA 151 MXN
470021-298
LaMotte® Dissolved Oxygen Sampler
Ingenious Design Ensures a Representative Sample
  • Calibrated line allows for samples at specific depths
  • Two-pound weight attached to the bottom of the sampling device
Just lower the clear, acrylic water sampler containing the 60 mL dissolved oxygen bottle to the desired depth. Water fills the bottle and overflows six times, ensuring an accurate, representative sample. The filled bottle can then be used to run a dissolved oxygen test, while the remaining water in the outer chambers of the sampler can be used for any other type of test you need. A nylon cord harness can attach to the calibrated line and is available separately. It comes with a water sampler, dissolved oxygen bottle, and sounding lead. Size of sampler: 3.5" square x 6.5" H.