CENCO® Lens and Mirror Clamp
CP72288-00
470110-468EA 66.99 MXN
470110-468
CENCO® Lens and Mirror Clamp
Unique, self-centering design accommodates a wide range of lens and mirror diameters.
  • Cenco quality and durability
This useful spring-loaded clamp securely holds lenses or mirrors from 2 to 10cm in diameter. It also can hold square or rectangular objects or gratings measuring 2 to 10cm on a side. It has a 1cm diameter support rod.