Set of Four Measuring Spoons
1045947MN
470149-714EA 60.8 MXN
470149-714
Set of Four Measuring Spoons
Cucharas
Four piece measuring spoon set: 1/4 tsp/1.25 mL, 1/2 tsp/2.5 mL, 1 tsp/5 mL, 1 Tbsp/15 mL.