Metric Micrometer, United Scientific Supplies
Proveedor: United Scientific Supplies
MCR025
470005-644EA 438.07 MXN
470005-644
Metric Micrometer, United Scientific Supplies
Calibradores
Excellent Quality
  • Precise
  • Forged steel frame
Ratchet stop ensures equal pressure on parts being measured.