Micropaleontology Slides, 1 mm Deep
: Avantor
Ideal for Temporary Mounts
Cardboard with 1 mm-deep cavities hold hard-to-see microfossils in a small area, highlighting them against a black background. Will last indefinitely when used with glass coverslips and aluminum slide holders. Size: 1" x 3". Packages of 100.