Modulated Visible Diode Laser
470006-428
: Avantor
- Description:Modulated visible diode laser
- Cat. no.:470006-428
- Supplier No.:ML 268
This laser does a lot of work!
- Easy to hold
- Includes manual
The 1.0 mW beam is a perfect pointer for A/V screens, walls, charts, or blackboards. It can be modulated to transmit voice or video and perform light speed measurement experiments (you must provide your own receiver). The compact size makes it easy to hold. It also features a 3-position power switch, a momentary pushbutton switch and aperture beam shutter. Power is supplied by rechargeable batteries or the external AC adaptor.