Precision pH Test Strips
Proveedor: Precision Laboratories
PH1013-1B-50 PH3060-1B-50 PH0114-1B-50 PH7010-12B-50 PH1114-1B-50 PH5090-1B-50 PH0015-12B-50
470150-880EA 174.73 MXN
470150-880 470150-918 470001-956 470150-806 470151-004 470150-922 470150-846
Precision pH Test Strips
Papeles de pruebas Tiras indicadoras de pH
The indicator on these strips won’t contaminate the solutions the strips are testing.
  • Vinyl-backed
  • Non-contaminating indicator
  • Fifty strips per package
Durable pH testing strips that are backed with vinyl.