Ribbed Glass Funnels, Variety Glass
Proveedor: VARIETY GLASS INC
695-7 695-5 695-3
470157-150CS 23747.17 MXN
470157-150 63025-014 470144-864
Ribbed Glass Funnels, Variety Glass
Embudos Embudos de filtración Embudos con filtro
Heavy glass with raised ribs inside.
  • Short beveled stem
  • Stem deeply grooved on outside to allow escape of air
These ribbed glass funnels are available with varied stem lengths (hand-cut), and are perfect for most laboratory applications.