SCIENCEWARE® Large Weighted Safety Shield, Thermo Scientific
Proveedor: BEL-ART PRODUCTS SE
H24962-0000_SE
470007-638EA 19975.82 MXN
470007-638
SCIENCEWARE® Large Weighted Safety Shield, Thermo Scientific
Pantallas Protectores de seguridad
Personnel are protected by a clear polycarbonate barrier with exceptional impact resistance and optical clarity.
  • 4.7mm (3/16") clear polycarbonate barrier
  • Exceptional impact resistance and optical clarity
Outside base dimensions are 50W x 17D cm (191/2 x 61/2"). This item ships oversize.
Certificaciones: Meets impact-resistance requirements of ANSI Z97.1.