SI Metric Desk Ruler
Proveedor: SPECTRUM EDUCATIONAL SUPPLIES
11340-A
470016-956EA 17.28 MXN
470016-956
SI Metric Desk Ruler
Reglas
One edge is graduated 0-30 cm with mm divisions. The other edge has three progressive scales: 100 mm, 10 cm, and 1 dm. On the reverse side is a metric SI unit and symbol chart. Made of plastic.